The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019589.2 | NP_062535.2 | 10 - 11 | 5468 | 68 |
XM_005267860.3 | XP_005267917.1 | 10 - 11 | 5512 | 68 | |
XM_011536966.2 | XP_011535268.1 | 11 - 12 | 4627 | 68 | |
XM_011536967.1 | XP_011535269.1 | 9 - 10 | 3394 | 68 | |
GenBank mRNA | AK090435.1 | - | 8 - 9 | 4224 | 68 |
AK297582.1 | - | 6 - 7 | 1039 | 68 | |
AL832365.1 | - | 7 - 8 | 4003 | 68 | |
BC007792.1 | - | 1 - 2 | 65 | 68 | |
L40403.1 | - | 4 - 5 | 571 | 68 |