Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SPECC1L
- Entrez Gene ID
-
23384
- Gene Name
- sperm antigen with calponin homology and coiled-coil domains 1 like
- Gene Aliases
-
CYTSA, GBBB2, OBLFC1
- Chromosome Location
-
Chr.22: 24270817 - 24417740 on Build GRCh38
- Gene Symbol
- SPECC1L-ADORA2A
- Entrez Gene ID
-
101730217
- Gene Name
- SPECC1L-ADORA2A readthrough (NMD candidate)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.22: 24270817 - 24442360 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_103546.1 |
|
17 - 18 |
3693 |
62 |
GenBank mRNA |
AB209022.1 |
- |
17 - 18 |
3612 |
62 |
View Full Product Details