The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270890.1 | NP_001257819.1 | 5 - 6 | 868 | 113 |
NM_017645.4 | NP_060115.3 | 5 - 6 | 868 | 113 | |
GenBank mRNA | AF533709.1 | - | 5 - 6 | 1050 | 113 |
AF537091.1 | - | 5 - 6 | 830 | 113 | |
AK022964.1 | - | 2 - 3 | 184 | 113 | |
AK056458.1 | - | 5 - 6 | 830 | 113 | |
AL832495.1 | - | 5 - 6 | 828 | 113 | |
BC026178.1 | - | 5 - 6 | 831 | 113 | |
BC065935.1 | - | 3 - 4 | 368 | 113 | |
BC111041.1 | - | 1 - 2 | 144 | 113 | |
BC114492.1 | - | 5 - 6 | 665 | 113 |