The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020935.2 | NP_065986.2 | 18 - 19 | 2313 | 71 |
XM_005246720.2 | XP_005246777.1 | 18 - 19 | 2385 | 71 | |
XM_005246721.3 | XP_005246778.1 | 18 - 19 | 2277 | 71 | |
XM_005246722.3 | XP_005246779.1 | 18 - 19 | 2317 | 71 | |
XM_011511538.2 | XP_011509840.1 | 17 - 18 | 2258 | 71 | |
XM_017004577.1 | XP_016860066.1 | 17 - 18 | 2173 | 71 | |
GenBank mRNA | AB046814.1 | - | 14 - 15 | 1758 | 71 |
AK095577.1 | - | 18 - 19 | 2271 | 71 | |
AK128583.1 | - | 18 - 19 | 2317 | 71 | |
AL832645.2 | - | 18 - 19 | 2312 | 71 | |
BC133007.1 | - | 18 - 19 | 2198 | 71 | |
BC133009.1 | - | 18 - 19 | 2243 | 71 | |
BC144249.1 | - | 18 - 19 | 2198 | 71 | |
BC144252.1 | - | 18 - 19 | 2238 | 71 | |
BX538024.1 | - | 18 - 19 | 2278 | 71 |