The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293274.1 | NP_001280203.1 | 25 - 26 | 3816 | 72 |
NM_020925.2 | NP_065976.2 | 26 - 27 | 3537 | 72 | |
XM_011541862.1 | XP_011540164.1 | 25 - 26 | 3812 | 72 | |
XM_011541863.1 | XP_011540165.1 | 26 - 27 | 3961 | 72 | |
GenBank mRNA | AB046793.1 | - | 24 - 25 | 3319 | 72 |
AK022571.1 | - | 13 - 14 | 1635 | 72 | |
AK024256.1 | - | 16 - 17 | 1962 | 72 | |
AK299361.1 | - | 12 - 13 | 1380 | 72 | |
AL833000.1 | - | 16 - 17 | 1986 | 72 | |
BC039301.1 | - | 26 - 27 | 3523 | 72 | |
BX537603.1 | - | 25 - 26 | 3815 | 72 |