Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- KANK1
- Entrez Gene ID
-
23189
- Gene Name
- KN motif and ankyrin repeat domains 1
- Gene Aliases
-
ANKRD15, CPSQ2, KANK
- Chromosome Location
-
Chr.9: 470294 - 746106 on Build GRCh38
View Full Product Details