Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CCDC14
- Entrez Gene ID
-
64770
- Gene Name
- coiled-coil domain containing 14
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 123887383 - 123961412 on Build GRCh38
View Full Product Details