The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011709.2 | NP_001011709.2 | 5 - 6 | 664 | 89 |
XM_011539276.1 | XP_011537578.1 | 5 - 6 | 697 | 89 | |
XM_011539277.1 | XP_011537579.1 | 3 - 4 | 288 | 89 | |
XM_011539278.1 | XP_011537580.1 | 5 - 6 | 568 | 89 | |
XM_011539279.1 | XP_011537581.1 | 4 - 5 | 542 | 89 | |
GenBank mRNA | AL833418.1 | - | 5 - 6 | 713 | 89 |
BC117224.1 | - | 5 - 6 | 664 | 89 |