Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLCCI1
Entrez Gene ID
113263
Gene Name
glucocorticoid induced 1
Gene Aliases
FAM117C, GCTR, GIG18, TSSN1
Chromosome Location
Chr.7: 7968743 - 8089080 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138426.3 NP_612435.1 6 - 7 1784 83
GenBank mRNA AL833694.1 - 2 - 3 279 83
BC050291.1 - 6 - 7 1735 83

View Full Product Details