The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198148.2 | NP_937791.2 | 4 - 5 | 980 | 69 |
XM_005269528.3 | XP_005269585.1 | 4 - 5 | 830 | 69 | |
XM_011539283.2 | XP_011537585.1 | 4 - 5 | 830 | 69 | |
XM_011539285.2 | XP_011537587.1 | 4 - 5 | 612 | 69 | |
XM_011539286.1 | XP_011537588.1 | 3 - 4 | 554 | 69 | |
XM_017015673.1 | XP_016871162.1 | 5 - 6 | 581 | 69 | |
GenBank mRNA | AK296365.1 | - | 4 - 5 | 387 | 69 |
AK304023.1 | - | 4 - 5 | 754 | 69 | |
AK304814.1 | - | 4 - 5 | 791 | 69 | |
AL833732.1 | - | 2 - 3 | 158 | 69 | |
AY358565.1 | - | 4 - 5 | 980 | 69 | |
BC027623.1 | - | 5 - 6 | 526 | 69 | |
BC036789.1 | - | 4 - 5 | 810 | 69 | |
BC137492.1 | - | 4 - 5 | 975 | 69 | |
BC146862.1 | - | 4 - 5 | 975 | 69 |