The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198148.2 | NP_937791.2 | 12 - 13 | 2245 | 66 |
XM_005269528.3 | XP_005269585.1 | 12 - 13 | 2095 | 66 | |
XM_011539283.2 | XP_011537585.1 | 12 - 13 | 2095 | 66 | |
XM_011539285.2 | XP_011537587.1 | 12 - 13 | 1877 | 66 | |
XM_011539286.1 | XP_011537588.1 | 11 - 12 | 1819 | 66 | |
XM_017015673.1 | XP_016871162.1 | 13 - 14 | 1846 | 66 | |
XM_017015674.1 | XP_016871163.1 | 6 - 7 | 1021 | 66 | |
GenBank mRNA | AK296365.1 | - | 12 - 13 | 1652 | 66 |
AK300506.1 | - | 8 - 9 | 1206 | 66 | |
AK304023.1 | - | 13 - 14 | 1944 | 66 | |
AK304814.1 | - | 12 - 13 | 2056 | 66 | |
AL833732.1 | - | 10 - 11 | 1423 | 66 | |
AY358565.1 | - | 12 - 13 | 2245 | 66 | |
BC027623.1 | - | 14 - 15 | 1968 | 66 | |
BC036789.1 | - | 12 - 13 | 2075 | 66 | |
BC137492.1 | - | 12 - 13 | 2240 | 66 | |
BC146862.1 | - | 12 - 13 | 2240 | 66 |