Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RCOR1
Entrez Gene ID
23186
Gene Name
REST corepressor 1
Gene Aliases
COREST, RCOR
Chromosome Location
Chr.14: 102592638 - 102730576 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015156.3 NP_055971.2 3 - 4 671 68
GenBank mRNA AF155595.1 - 3 - 4 434 68
BC051003.1 - 3 - 4 669 68
BC064495.1 - 3 - 4 669 68
D31888.1 - 3 - 4 176 68

View Full Product Details