Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 31058280-31058777 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WRN
Entrez Gene ID
7486
Gene Name
Werner syndrome RecQ like helicase
Gene Aliases
RECQ3, RECQL2, RECQL3
UniGene
Hs.632050
Gene Chromosome Location
Chr.8: 31033262-31173761 on Build GRCh38
Transcripts
NM_000553.4 XM_011544639.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7397942 c.95A>G p.K32R Chr.8: 31058542-31058542 on Build GRCh38
750650 c.96G>T p.K32N Chr.8: 31058543-31058543 on Build GRCh38
3731362 c.8delA p.K5fs*15 Chr.8: 31058455-31058455 on Build GRCh38
1099217 c.38G>A p.R13Q Chr.8: 31058485-31058485 on Build GRCh38
2958600 c.33G>A p.Q11Q Chr.8: 31058480-31058480 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759713090 Chr.8: 31058562-31058562 on Build GRCh38
rs200352487 Chr.8: 31058580-31058580 on Build GRCh38
rs763753170 Chr.8: 31058422-31058422 on Build GRCh38
rs1049484172 Chr.8: 31058735-31058735 on Build GRCh38
rs3087430 Chr.8: 31058599-31058599 on Build GRCh38
rs1001110767 Chr.8: 31058363-31058363 on Build GRCh38
rs754627643 Chr.8: 31058520-31058520 on Build GRCh38
rs971153103 Chr.8: 31058743-31058743 on Build GRCh38
rs370014019 Chr.8: 31058411-31058411 on Build GRCh38
rs888468405 Chr.8: 31058475-31058475 on Build GRCh38
rs544479890 Chr.8: 31058651-31058651 on Build GRCh38
rs778355874 Chr.8: 31058526-31058526 on Build GRCh38
rs916991533 Chr.8: 31058340-31058340 on Build GRCh38
rs971099690 Chr.8: 31058607-31058607 on Build GRCh38
rs187869929 Chr.8: 31058397-31058397 on Build GRCh38
rs538314935 Chr.8: 31058529-31058529 on Build GRCh38
rs753595348 Chr.8: 31058424-31058424 on Build GRCh38
rs756255418 Chr.8: 31058480-31058480 on Build GRCh38
rs776465139 Chr.8: 31058570-31058570 on Build GRCh38
rs755891046 Chr.8: 31058543-31058543 on Build GRCh38
rs973997819 Chr.8: 31058551-31058551 on Build GRCh38
rs749390862 Chr.8: 31058547-31058547 on Build GRCh38
rs1036117223 Chr.8: 31058355-31058355 on Build GRCh38
rs770866944 Chr.8: 31058549-31058549 on Build GRCh38
rs3087420 Chr.8: 31058489-31058489 on Build GRCh38
rs919114323 Chr.8: 31058668-31058668 on Build GRCh38
rs375337235 Chr.8: 31058573-31058573 on Build GRCh38
rs538425690 Chr.8: 31058337-31058337 on Build GRCh38
rs2737316 Chr.8: 31058569-31058569 on Build GRCh38
rs949896878 Chr.8: 31058349-31058349 on Build GRCh38
rs1034292863 Chr.8: 31058380-31058380 on Build GRCh38
rs779625263 Chr.8: 31058580-31058580 on Build GRCh38
rs945442735 Chr.8: 31058583-31058583 on Build GRCh38
rs1025560768 Chr.8: 31058718-31058718 on Build GRCh38
rs774377745 Chr.8: 31058402-31058402 on Build GRCh38
rs535823414 Chr.8: 31058724-31058724 on Build GRCh38
rs781459943 Chr.8: 31058521-31058521 on Build GRCh38
rs373503267 Chr.8: 31058484-31058484 on Build GRCh38
rs202148988 Chr.8: 31058463-31058463 on Build GRCh38
rs201284343 Chr.8: 31058574-31058574 on Build GRCh38
rs565466453 Chr.8: 31058620-31058620 on Build GRCh38
rs904146243 Chr.8: 31058320-31058320 on Build GRCh38
rs1006884728 Chr.8: 31058485-31058485 on Build GRCh38
rs761784221 Chr.8: 31058459-31058459 on Build GRCh38
rs34477820 Chr.8: 31058542-31058542 on Build GRCh38
rs757791580 Chr.8: 31058472-31058472 on Build GRCh38
rs560980155 Chr.8: 31058678-31058678 on Build GRCh38
rs150524008 Chr.8: 31058522-31058522 on Build GRCh38
rs903843755 Chr.8: 31058605-31058605 on Build GRCh38
rs1033260656 Chr.8: 31058576-31058576 on Build GRCh38
rs1013714904 Chr.8: 31058700-31058700 on Build GRCh38
rs749073678 Chr.8: 31058564-31058564 on Build GRCh38
rs878854133 Chr.8: 31058462-31058462 on Build GRCh38
rs1024022898 Chr.8: 31058448-31058448 on Build GRCh38
rs193031348 Chr.8: 31058400-31058400 on Build GRCh38
rs555946689 Chr.8: 31058638-31058638 on Build GRCh38
rs550926459 Chr.8: 31058524-31058524 on Build GRCh38
rs530000192 Chr.8: 31058694-31058694 on Build GRCh38
rs531155309 Chr.8: 31058635-31058635 on Build GRCh38
rs377656120 Chr.8: 31058552-31058552 on Build GRCh38

View Full Product Details