Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 31058963-31059471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WRN
Entrez Gene ID
7486
Gene Name
Werner syndrome RecQ like helicase
Gene Aliases
RECQ3, RECQL2, RECQL3
UniGene
Hs.632050
Gene Chromosome Location
Chr.8: 31033262-31173761 on Build GRCh38
Transcripts
NM_000553.4 XM_011544639.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1673844 c.142G>T p.E48* Chr.8: 31059198-31059198 on Build GRCh38
286547 c.172G>A p.D58N Chr.8: 31059228-31059228 on Build GRCh38
5611173 c.97G>A p.A33T Chr.8: 31059153-31059153 on Build GRCh38
3648342 c.156C>T p.S52S Chr.8: 31059212-31059212 on Build GRCh38
1699943 c.104T>C p.V35A Chr.8: 31059160-31059160 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868599739 Chr.8: 31059266-31059266 on Build GRCh38
rs76077884 Chr.8: 31059394-31059394 on Build GRCh38
rs780988648 Chr.8: 31059152-31059152 on Build GRCh38
rs923173679 Chr.8: 31059387-31059387 on Build GRCh38
rs954022892 Chr.8: 31059158-31059158 on Build GRCh38
rs139775895 Chr.8: 31059186-31059186 on Build GRCh38
rs750315075 Chr.8: 31059114-31059114 on Build GRCh38
rs1003935557 Chr.8: 31059060-31059060 on Build GRCh38
rs866361372 Chr.8: 31059307-31059307 on Build GRCh38
rs370347185 Chr.8: 31059130-31059130 on Build GRCh38
rs1019515522 Chr.8: 31059427-31059427 on Build GRCh38
rs1046605858 Chr.8: 31059312-31059312 on Build GRCh38
rs1024495075 Chr.8: 31059262-31059262 on Build GRCh38
rs762851957 Chr.8: 31059216-31059216 on Build GRCh38
rs775635001 Chr.8: 31059199-31059199 on Build GRCh38
rs951805442 Chr.8: 31059335-31059335 on Build GRCh38
rs940826673 Chr.8: 31059362-31059362 on Build GRCh38
rs372306347 Chr.8: 31059277-31059277 on Build GRCh38
rs754208761 Chr.8: 31059270-31059270 on Build GRCh38
rs949684941 Chr.8: 31059311-31059311 on Build GRCh38
rs534118637 Chr.8: 31059091-31059091 on Build GRCh38
rs748942438 Chr.8: 31059122-31059123 on Build GRCh38
rs745937507 Chr.8: 31059305-31059305 on Build GRCh38
rs762558928 Chr.8: 31059126-31059126 on Build GRCh38
rs201800431 Chr.8: 31059170-31059170 on Build GRCh38
rs943237754 Chr.8: 31059062-31059062 on Build GRCh38
rs753114305 Chr.8: 31059155-31059155 on Build GRCh38
rs993090076 Chr.8: 31059315-31059315 on Build GRCh38
rs962699213 Chr.8: 31059095-31059095 on Build GRCh38
rs747102789 Chr.8: 31059322-31059322 on Build GRCh38
rs759022881 Chr.8: 31059232-31059232 on Build GRCh38
rs188604526 Chr.8: 31059364-31059364 on Build GRCh38
rs201590676 Chr.8: 31059185-31059185 on Build GRCh38
rs768226589 Chr.8: 31059191-31059193 on Build GRCh38
rs11574178 Chr.8: 31059371-31059371 on Build GRCh38
rs922619165 Chr.8: 31059107-31059107 on Build GRCh38
rs764589321 Chr.8: 31059257-31059257 on Build GRCh38
rs773803207 Chr.8: 31059228-31059228 on Build GRCh38
rs373806031 Chr.8: 31059227-31059227 on Build GRCh38
rs772246871 Chr.8: 31059193-31059193 on Build GRCh38
rs545879766 Chr.8: 31059359-31059359 on Build GRCh38
rs757761103 Chr.8: 31059273-31059273 on Build GRCh38
rs1028019506 Chr.8: 31059372-31059372 on Build GRCh38
rs772963859 Chr.8: 31059213-31059213 on Build GRCh38
rs36054800 Chr.8: 31059110-31059111 on Build GRCh38
rs898286356 Chr.8: 31059369-31059369 on Build GRCh38
rs925324252 Chr.8: 31059222-31059222 on Build GRCh38
rs34084741 Chr.8: 31059163-31059163 on Build GRCh38
rs995146854 Chr.8: 31059124-31059124 on Build GRCh38
rs538178496 Chr.8: 31059160-31059160 on Build GRCh38
rs775488767 Chr.8: 31059395-31059395 on Build GRCh38
rs756575284 Chr.8: 31059253-31059253 on Build GRCh38
rs141495269 Chr.8: 31059162-31059162 on Build GRCh38
rs752345825 Chr.8: 31059238-31059238 on Build GRCh38
rs751059046 Chr.8: 31059141-31059141 on Build GRCh38
rs576906241 Chr.8: 31059326-31059326 on Build GRCh38
rs754646679 Chr.8: 31059143-31059143 on Build GRCh38
rs145959045 Chr.8: 31059200-31059200 on Build GRCh38
rs758424926 Chr.8: 31059309-31059309 on Build GRCh38
rs765851196 Chr.8: 31059127-31059127 on Build GRCh38
rs767288688 Chr.8: 31059234-31059234 on Build GRCh38
rs771433734 Chr.8: 31059167-31059167 on Build GRCh38
rs587778749 Chr.8: 31059201-31059201 on Build GRCh38

View Full Product Details