Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UGT8
Entrez Gene ID
7368
Gene Name
UDP glycosyltransferase 8
Gene Aliases
CGT, UGT4
Chromosome Location
Chr.4: 114597789 - 114687914 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128174.2 NP_001121646.1 3 - 4 1540 157
NM_001322112.1 NP_001309041.1 4 - 5 1643 157
NM_001322113.1 NP_001309042.1 3 - 4 1623 157
NM_001322114.1 NP_001309043.1 3 - 4 1469 157
NM_003360.4 NP_003351.2 2 - 3 1479 157
GenBank mRNA AK127970.1 - 3 - 4 1470 157
AK225763.1 - 3 - 4 1133 157
BC075069.2 - 3 - 4 1012 157
U30930.1 - 2 - 3 1480 157
U62899.1 - 3 - 4 1191 157

View Full Product Details