The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000260.3 | NP_000251.3 | 24 - 25 | 3381 | 76 |
NM_001127179.2 | NP_001120651.2 | 24 - 25 | 3381 | 76 | |
NM_001127180.1 | NP_001120652.1 | 24 - 25 | 3375 | 76 | |
XM_011545044.2 | XP_011543346.1 | 24 - 25 | 3453 | 76 | |
XM_011545046.2 | XP_011543348.2 | 23 - 24 | 3214 | 76 | |
XM_011545050.2 | XP_011543352.1 | 21 - 22 | 3068 | 76 | |
XM_017017778.1 | XP_016873267.1 | 23 - 24 | 3214 | 76 | |
XM_017017779.1 | XP_016873268.1 | 23 - 24 | 3214 | 76 | |
XM_017017780.1 | XP_016873269.1 | 23 - 24 | 3214 | 76 | |
XM_017017781.1 | XP_016873270.1 | 23 - 24 | 3214 | 76 | |
XM_017017782.1 | XP_016873271.1 | 23 - 24 | 3214 | 76 | |
XM_017017783.1 | XP_016873272.1 | 23 - 24 | 3214 | 76 | |
XM_017017784.1 | XP_016873273.1 | 23 - 24 | 3214 | 76 | |
XM_017017785.1 | XP_016873274.1 | 21 - 22 | 2983 | 76 | |
XM_017017786.1 | XP_016873275.1 | 23 - 24 | 3214 | 76 | |
XM_017017787.1 | XP_016873276.1 | 23 - 24 | 3214 | 76 | |
XM_017017788.1 | XP_016873277.1 | 23 - 24 | 3214 | 76 | |
GenBank mRNA | AB209629.1 | - | 4 - 5 | 652 | 76 |
AB290181.1 | - | 4 - 5 | 652 | 76 | |
BC023212.1 | - | 9 - 10 | 1389 | 76 | |
BC144519.1 | - | 25 - 26 | 3423 | 76 | |
U34227.1 | - | 24 - 25 | 3376 | 76 | |
U39226.1 | - | 24 - 25 | 3381 | 76 | |
U55208.1 | - | 24 - 25 | 3376 | 76 | |
U55209.1 | - | 24 - 25 | 3376 | 76 |