Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM69
Entrez Gene ID
51249
Gene Name
transmembrane protein 69
Gene Aliases
C1orf154
Chromosome Location
Chr.1: 45688175 - 45694436 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016486.3 NP_057570.2 1 - 2 103 63
GenBank mRNA AF151063.1 - 1 - 2 83 63
AK024682.1 - 1 - 2 97 63
BC013608.1 - 1 - 2 85 63
BC034147.1 - 1 - 2 83 63
BC046130.1 - 1 - 2 66 63
BC064539.1 - 1 - 2 86 63
BC121117.2 - 1 - 2 45 63
BC133692.1 - 1 - 2 44 63

View Full Product Details