The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286577.1 | NP_001273506.1 | 29 - 30 | 5886 | 64 |
NM_015531.5 | NP_056346.3 | 29 - 30 | 5886 | 64 | |
XM_011544904.1 | XP_011543206.1 | 29 - 30 | 5785 | 64 | |
XM_011544905.1 | XP_011543207.1 | 29 - 30 | 5785 | 64 | |
XM_017017510.1 | XP_016872999.1 | 29 - 30 | 5785 | 64 | |
XM_017017511.1 | XP_016873000.1 | 28 - 29 | 5711 | 64 | |
XM_017017512.1 | XP_016873001.1 | 28 - 29 | 5676 | 64 | |
XM_017017513.1 | XP_016873002.1 | 19 - 20 | 3882 | 64 | |
XM_017017515.1 | XP_016873004.1 | 15 - 16 | 3435 | 64 | |
XM_017017516.1 | XP_016873005.1 | 15 - 16 | 3005 | 64 | |
GenBank mRNA | AB231764.1 | - | 29 - 30 | 5700 | 64 |
AK125894.1 | - | 16 - 17 | 3547 | 64 | |
AL833903.1 | - | 10 - 11 | 2083 | 64 | |
BC110508.2 | - | 16 - 17 | 3515 | 64 | |
CD369754.1 | - | 2 - 3 | 64 |