Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EHBP1
Entrez Gene ID
23301
Gene Name
EH domain binding protein 1
Gene Aliases
HPC12, NACSIN
Chromosome Location
Chr.2: 62673851 - 63046487 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142614.1 NP_001136086.1 20 - 21 3603 118
NM_001142615.2 NP_001136087.1 20 - 21 3728 118
NM_001142616.1 NP_001136088.1 20 - 21 3682 118
NM_015252.3 NP_056067.2 22 - 23 3895 118
XM_005264225.2 XP_005264282.1 22 - 23 3793 118
XM_005264226.2 XP_005264283.1 22 - 23 4150 118
XM_005264227.1 XP_005264284.1 21 - 22 3749 118
XM_005264228.2 XP_005264285.1 22 - 23 3008 118
XM_006711971.2 XP_006712034.1 21 - 22 3746 118
XM_011532713.2 XP_011531015.1 22 - 23 3936 118
XM_011532714.2 XP_011531016.1 22 - 23 3785 118
XM_011532715.2 XP_011531017.1 22 - 23 3761 118
XM_011532716.2 XP_011531018.1 22 - 23 3851 118
XM_011532717.1 XP_011531019.1 21 - 22 3743 118
XM_011532718.2 XP_011531020.1 21 - 22 3515 118
XM_017003643.1 XP_016859132.1 22 - 23 3751 118
XM_017003644.1 XP_016859133.1 22 - 23 3728 118
XM_017003645.1 XP_016859134.1 22 - 23 3790 118
XM_017003646.1 XP_016859135.1 22 - 23 3853 118
XM_017003647.1 XP_016859136.1 21 - 22 3688 118
XM_017003648.1 XP_016859137.1 21 - 22 3684 118
XM_017003649.1 XP_016859138.1 21 - 22 3763 118
XM_017003650.1 XP_016859139.1 21 - 22 3685 118
XM_017003651.1 XP_016859140.1 20 - 21 3580 118
XM_017003652.1 XP_016859141.1 20 - 21 3638 118
XM_017003653.1 XP_016859142.1 20 - 21 3566 118
XM_017003655.1 XP_016859144.1 20 - 21 2795 118
GenBank mRNA AB020710.1 - 14 - 15 2608 118
AK000986.1 - 6 - 7 733 118
AK292545.1 - 20 - 21 3728 118
AK309125.1 - 7 - 8 816 118
AL833968.1 - 22 - 23 3761 118
AY331186.1 - 22 - 23 3790 118
AY531390.1 - 20 - 21 3308 118
BC067215.1 - 20 - 21 3536 118

View Full Product Details