The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006828.3 | NP_006819.2 | 23 - 24 | 4091 | 143 |
XM_011535394.2 | XP_011533696.1 | 23 - 24 | 4101 | 143 | |
XM_011535395.2 | XP_011533697.1 | 24 - 25 | 4326 | 143 | |
XM_011535396.2 | XP_011533698.1 | 22 - 23 | 3935 | 143 | |
XM_017010205.1 | XP_016865694.1 | 24 - 25 | 4347 | 143 | |
XM_017010206.1 | XP_016865695.1 | 24 - 25 | 4125 | 143 | |
GenBank mRNA | AJ223948.1 | - | 6 - 7 | 930 | 143 |
AL834463.1 | - | 22 - 23 | 3776 | 143 | |
AY013288.1 | - | 19 - 20 | 2910 | 143 |