Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DHX29
Entrez Gene ID
54505
Gene Name
DEAH-box helicase 29
Gene Aliases
DDX29
Chromosome Location
Chr.5: 55256245 - 55307693 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019030.2 NP_061903.2 11 - 12 2084 91
GenBank mRNA AL079292.1 - 1 - 2 286 91
AL834496.1 - 1 - 2 351 91
AY036974.1 - 11 - 12 2065 91
BC056219.1 - 11 - 12 2061 91
BX648101.1 - 10 - 11 1732 91
BX648269.1 - 11 - 12 1799 91
BX648688.1 - 4 - 5 986 91

View Full Product Details