The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019030.2 | NP_061903.2 | 24 - 25 | 3955 | 138 |
XM_006714653.2 | XP_006714716.1 | 24 - 25 | 3792 | 138 | |
XM_017009588.1 | XP_016865077.1 | 24 - 25 | 4032 | 138 | |
GenBank mRNA | AF070639.1 | - | 3 - 4 | 482 | 138 |
AK025145.1 | - | 11 - 12 | 1425 | 138 | |
AL079292.1 | - | 14 - 15 | 2157 | 138 | |
AL834496.1 | - | 14 - 15 | 2222 | 138 | |
AY036974.1 | - | 24 - 25 | 3936 | 138 | |
BC056219.1 | - | 24 - 25 | 3932 | 138 | |
BX648101.1 | - | 23 - 24 | 3603 | 138 | |
BX648269.1 | - | 24 - 25 | 3670 | 138 | |
BX648688.1 | - | 17 - 18 | 2857 | 138 | |
BX649135.1 | - | 24 - 25 | 4044 | 138 |