The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153447.4 | NP_703148.4 | 6 - 7 | 678 | 85 |
GenBank mRNA | AY054986.1 | - | 6 - 7 | 678 | 85 |
AY154460.1 | - | 6 - 7 | 678 | 85 |