The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138477.2 | NP_612486.2 | 23 - 24 | 3211 | 109 |
XM_005254176.4 | XP_005254233.1 | 23 - 24 | 3126 | 109 | |
XM_011521270.2 | XP_011519572.1 | 23 - 24 | 3151 | 109 | |
XM_011521271.2 | XP_011519573.1 | 23 - 24 | 3148 | 109 | |
XM_011521274.2 | XP_011519576.1 | 20 - 21 | 2197 | 109 | |
GenBank mRNA | AF525398.1 | - | 23 - 24 | 3208 | 109 |
BC001092.2 | - | 15 - 16 | 1666 | 109 | |
BC008333.1 | - | 15 - 16 | 1666 | 109 | |
BC008334.1 | - | 15 - 16 | 1666 | 109 | |
BC052568.1 | - | 23 - 24 | 3099 | 109 | |
BC066640.1 | - | 23 - 24 | 3099 | 109 |