Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNTRL
Entrez Gene ID
11064
Gene Name
centriolin
Gene Aliases
CEP1, CEP110, FAN, bA165P4.1
Chromosome Location
Chr.9: 121074778 - 121177610 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007018.4 NP_008949.4 12 - 13 2062 64
XM_005251679.3 XP_005251736.1 14 - 15 2334 64
XM_006716940.2 XP_006717003.1 14 - 15 2334 64
XM_006716942.1 XP_006717005.1 3 - 4 563 64
XM_006716944.3 XP_006717007.1 14 - 15 2334 64
XM_011518166.1 XP_011516468.1 14 - 15 2334 64
XM_011518167.1 XP_011516469.1 13 - 14 2175 64
XM_011518168.1 XP_011516470.1 14 - 15 2259 64
XM_011518169.2 XP_011516471.1 13 - 14 2209 64
XM_011518172.2 XP_011516474.1 14 - 15 2334 64
XM_011518173.1 XP_011516475.1 2 - 3 381 64
XM_017014223.1 XP_016869712.1 14 - 15 2334 64
XM_017014224.1 XP_016869713.1 14 - 15 2334 64
XM_017014225.1 XP_016869714.1 12 - 13 2020 64
XM_017014226.1 XP_016869715.1 3 - 4 563 64
XM_017014227.1 XP_016869716.1 13 - 14 2352 64
XM_017014228.1 XP_016869717.1 13 - 14 2208 64
XM_017014229.1 XP_016869718.1 14 - 15 2334 64
XM_017014230.1 XP_016869719.1 14 - 15 2334 64
GenBank mRNA AF513978.1 - 12 - 13 2031 64
AK074079.1 - 3 - 4 453 64
AK090600.1 - 3 - 4 532 64
AK097636.1 - 6 - 7 939 64
AK128098.1 - 10 - 11 1576 64
AY651261.1 - 5 - 6 734 64
BC002932.2 - 3 - 4 517 64
BC137284.1 - 12 - 13 2055 64
BC137286.1 - 12 - 13 2054 64

View Full Product Details