Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LMOD2
Entrez Gene ID
442721
Gene Name
leiomodin 2
Gene Aliases
C-LMOD, CLMOD
Chromosome Location
Chr.7: 123655807 - 123664093 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207163.1 NP_997046.1 1 - 2 425 103
GenBank mRNA BC017911.1 - 1 - 2 366 103

View Full Product Details