The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145166.1 | NP_001138638.1 | 4 - 5 | 593 | 69 |
NM_001145167.1 | NP_001138639.1 | 5 - 6 | 643 | 69 | |
NM_005788.3 | NP_005779.1 | 6 - 7 | 779 | 69 | |
XM_011519836.2 | XP_011518138.1 | 3 - 4 | 756 | 69 | |
XM_017017081.1 | XP_016872570.1 | 5 - 6 | 515 | 69 | |
GenBank mRNA | AF059531.1 | - | 5 - 6 | 506 | 69 |
AK129984.1 | - | - | 697 | 69 | |
AK298185.1 | - | 5 - 6 | 457 | 69 | |
AK300591.1 | - | 4 - 5 | 407 | 69 | |
AK301449.1 | - | 6 - 7 | 611 | 69 | |
AK307776.1 | - | 2 - 3 | 238 | 69 | |
BC019339.1 | - | 5 - 6 | 615 | 69 | |
BC064831.1 | - | 6 - 7 | 754 | 69 |