The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199159.1 | NP_001186088.1 | 6 - 7 | 1493 | 91 |
NM_153370.2 | NP_699201.2 | 5 - 6 | 1598 | 91 | |
XM_011514375.2 | XP_011512677.1 | 5 - 6 | 1427 | 91 | |
GenBank mRNA | AK075470.1 | - | 5 - 6 | 1293 | 91 |
AK302193.1 | - | 4 - 5 | 1101 | 91 | |
AK303928.1 | - | 4 - 5 | 1247 | 91 | |
AK312785.1 | - | 5 - 6 | 1291 | 91 | |
BC022399.1 | - | 5 - 6 | 1293 | 91 | |
BC035634.1 | - | 5 - 6 | 1598 | 91 |