The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002471.3 | NP_002462.2 | 15 - 16 | 1960 | 89 |
GenBank mRNA | BC132667.1 | - | 14 - 15 | 1919 | 89 |
D00943.1 | - | 13 - 14 | 1889 | 89 |