The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015045.3 | NP_055860.1 | 3 - 4 | 1965 | 73 |
XM_017015980.1 | XP_016871469.1 | 3 - 4 | 1958 | 73 | |
GenBank mRNA | AB065003.1 | - | 3 - 4 | 1968 | 73 |
AK290138.1 | - | 3 - 4 | 1926 | 73 | |
BC150269.1 | - | 4 - 5 | 1816 | 73 | |
D87450.1 | - | 4 - 5 | 1816 | 73 |