Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBXN4
Entrez Gene ID
23190
Gene Name
UBX domain protein 4
Gene Aliases
UBXD2, UBXDC1, erasin
Chromosome Location
Chr.2: 135741619 - 135785063 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014607.3 NP_055422.1 5 - 6 818 67
GenBank mRNA AK026971.1 - 5 - 6 441 67
AK292829.1 - 5 - 6 511 67
AK304096.1 - 5 - 6 528 67
BC020806.1 - 4 - 5 1160 67
BC035594.2 - 6 - 7 650 67
BX647662.1 - 5 - 6 610 67
D87684.1 - 5 - 6 571 67
EF199840.1 - 5 - 6 635 67

View Full Product Details