Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
INSRR
Entrez Gene ID
3645
Gene Name
insulin receptor related receptor
Gene Aliases
IRR
Chromosome Location
Chr.1: 156840873 - 156858920 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014215.2 NP_055030.1 20 - 21 3830 82
GenBank mRNA J05046.1 - 19 - 20 3446 82

View Full Product Details