The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017617.4 | NP_060087.3 | 23 - 24 | 3902 | 83 |
XM_011518717.2 | XP_011517019.2 | 20 - 21 | 3194 | 83 | |
GenBank mRNA | AB209873.1 | - | 15 - 16 | 2439 | 83 |
AF308602.1 | - | 23 - 24 | 3905 | 83 | |
M73980.1 | - | 23 - 24 | 3905 | 83 |