The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160102.1 | NP_001153574.1 | 4 - 5 | 636 | 128 |
NM_006151.2 | NP_006142.1 | 6 - 7 | 885 | 128 | |
NR_027647.1 | 5 - 6 | 797 | 128 | ||
XM_011524808.2 | XP_011523110.1 | 6 - 7 | 761 | 128 | |
GenBank mRNA | AK131496.1 | - | 4 - 5 | 1936 | 128 |
AK300653.1 | - | 4 - 5 | 361 | 128 | |
AK303903.1 | - | 5 - 6 | 632 | 128 | |
AK308378.1 | - | 6 - 7 | 610 | 128 | |
AK310965.1 | - | 6 - 7 | 640 | 128 | |
BC107166.1 | - | 6 - 7 | 756 | 128 | |
BC107167.1 | - | 6 - 7 | 756 | 128 | |
BX538344.1 | - | 2 - 3 | 206 | 128 | |
BX648309.1 | - | 6 - 7 | 723 | 128 | |
EF579964.1 | - | 4 - 5 | 358 | 128 | |
U39573.1 | - | 6 - 7 | 650 | 128 |