The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032242.3 | NP_115618.3 | 8 - 9 | 2115 | 53 |
XM_011512908.2 | XP_011511210.1 | 9 - 10 | 2217 | 53 | |
XM_011512909.1 | XP_011511211.1 | 9 - 10 | 2810 | 53 | |
GenBank mRNA | X87832.2 | - | 8 - 9 | 2050 | 53 |