The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000226.3 | NP_000217.2 | 1 - 2 | 706 | 94 |
GenBank mRNA | AB001594.1 | - | 1 - 2 | 203 | 94 |
BC121170.2 | - | 2 - 3 | 172 | 94 | |
S69510.1 | - | 1 - 2 | 703 | 94 | |
Z29074.1 | - | 1 - 2 | 706 | 94 |