The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024916.3 | NP_001020087.2 | 9 - 10 | 811 | 81 |
NM_001286835.1 | NP_001273764.1 | 13 - 14 | 2063 | 81 | |
XM_005272748.1 | XP_005272805.1 | 11 - 12 | 915 | 81 | |
XM_005272749.1 | XP_005272806.1 | 10 - 11 | 858 | 81 | |
XM_005272750.1 | XP_005272807.1 | 10 - 11 | 855 | 81 | |
XM_005272751.1 | XP_005272808.1 | 10 - 11 | 828 | 81 | |
XM_006716914.1 | XP_006716977.1 | 11 - 12 | 915 | 81 | |
XM_017014455.1 | XP_016869944.1 | 10 - 11 | 828 | 81 | |
XM_017014456.1 | XP_016869945.1 | 12 - 13 | 846 | 81 | |
XM_017014457.1 | XP_016869946.1 | 11 - 12 | 759 | 81 | |
XM_017014459.1 | XP_016869948.1 | 13 - 14 | 1094 | 81 | |
XM_017014461.1 | XP_016869950.1 | 12 - 13 | 1007 | 81 | |
GenBank mRNA | AK097639.1 | - | 11 - 12 | 1448 | 81 |
AK292585.1 | - | 12 - 13 | 983 | 81 | |
BC067803.1 | - | 13 - 14 | 2001 | 81 | |
BC082271.1 | - | 9 - 10 | 707 | 81 | |
BM711778.1 | - | 2 - 3 | 73 | 81 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145355.1 | NP_001138827.1 | 12 - 13 | 1016 | 81 |
NM_001145356.1 | NP_001138828.1 | 10 - 11 | 861 | 81 | |
NM_018491.3 | NP_060961.3 | 11 - 12 | 918 | 81 | |
XM_005251513.4 | XP_005251570.1 | 10 - 11 | 866 | 81 | |
XM_005251514.1 | XP_005251571.1 | 10 - 11 | 831 | 81 | |
XM_005251515.1 | XP_005251572.1 | 9 - 10 | 774 | 81 | |
XM_011517961.2 | XP_011516263.1 | 11 - 12 | 953 | 81 | |
XM_011517962.2 | XP_011516264.1 | 11 - 12 | 918 | 81 | |
XM_011517963.1 | XP_011516265.1 | 8 - 9 | 456 | 81 | |
GenBank mRNA | AF212253.1 | - | 10 - 11 | 829 | 81 |
AF257330.2 | - | 11 - 12 | 876 | 81 | |
AK290648.1 | - | 12 - 13 | 988 | 81 | |
AK297292.1 | - | 11 - 12 | 900 | 81 | |
AY343911.1 | - | 11 - 12 | 875 | 81 | |
BC005996.1 | - | 11 - 12 | 918 | 81 | |
BC009573.1 | - | 12 - 13 | 969 | 81 | |
BC013432.1 | - | 11 - 12 | 889 | 81 | |
BC132759.1 | - | 11 - 12 | 852 | 81 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172003.3 | NP_742000.1 | 11 - 12 | 990 | 81 |
XM_005263600.2 | XP_005263657.1 | 10 - 11 | 933 | 81 | |
XM_005263602.2 | XP_005263659.1 | 10 - 11 | 903 | 81 | |
XM_005263603.2 | XP_005263660.1 | 12 - 13 | 1024 | 81 | |
XM_005263604.2 | XP_005263661.1 | 9 - 10 | 846 | 81 | |
XM_005263605.3 | XP_005263662.1 | 11 - 12 | 968 | 81 | |
XM_011510667.1 | XP_011508969.1 | 11 - 12 | 990 | 81 | |
XM_011510668.2 | XP_011508970.1 | 8 - 9 | 550 | 81 | |
XM_011510669.2 | XP_011508971.1 | 12 - 13 | 1034 | 81 | |
XM_017003408.1 | XP_016858897.1 | 13 - 14 | 1154 | 81 | |
GenBank mRNA | AF452722.1 | - | 11 - 12 | 846 | 81 |
AK124867.1 | - | 2 - 3 | 2555 | 81 | |
AK225251.1 | - | 12 - 13 | 1004 | 81 | |
AK290072.1 | - | 11 - 12 | 990 | 81 | |
AY343910.1 | - | 11 - 12 | 875 | 81 | |
BC120991.1 | - | 11 - 12 | 861 | 81 | |
BC120992.1 | - | 11 - 12 | 861 | 81 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291821.1 | NP_001278750.1 | 10 - 11 | 839 | 81 |
NM_201453.3 | NP_958861.2 | 11 - 12 | 899 | 81 | |
XM_005277637.2 | XP_005277694.1 | 10 - 11 | 921 | 81 | |
XM_005277639.2 | XP_005277696.1 | 10 - 11 | 891 | 81 | |
XM_005277640.2 | XP_005277697.1 | 9 - 10 | 834 | 81 | |
XM_006717116.1 | XP_006717179.1 | 11 - 12 | 978 | 81 | |
XM_011518706.2 | XP_011517008.1 | 6 - 7 | 429 | 81 | |
XM_017014730.1 | XP_016870219.1 | 9 - 10 | 831 | 81 | |
XM_017014731.1 | XP_016870220.1 | 11 - 12 | 3350 | 81 | |
XM_017014732.1 | XP_016870221.1 | 5 - 6 | 342 | 81 | |
XM_017014733.1 | XP_016870222.1 | 6 - 7 | 5649 | 81 | |
GenBank mRNA | AK297912.1 | - | 10 - 11 | 801 | 81 |
AY344633.1 | - | 11 - 12 | 875 | 81 |