Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TBC1D3E
- Entrez Gene ID
-
102723859
- Gene Name
- TBC1 domain family member 3E
- Gene Aliases
-
TBC1DE3
- Chromosome Location
-
Chr.17: 38119293 - 38138868 on Build GRCh38
- Gene Symbol
- LOC101060389
- Entrez Gene ID
-
101060389
- Gene Name
- TBC1 domain family member-like
- Gene Aliases
-
TBC1D3D
- Chromosome Location
-
Chr.17: 38004033 - 38014908 on Build GRCh38
- Gene Symbol
- TBC1D3
- Entrez Gene ID
-
729873
- Gene Name
- TBC1 domain family member 3
- Gene Aliases
-
PRC17, TBC1D3A, TBC1D3F
- Chromosome Location
-
Chr.17: 38181476 - 38192555 on Build GRCh38
- Gene Symbol
- TBC1D3C
- Entrez Gene ID
-
414060
- Gene Name
- TBC1 domain family member 3C
- Gene Aliases
-
TBC1D3D
- Chromosome Location
-
Chr.17: 38057693 - 38068634 on Build GRCh38
- Gene Symbol
- TBC1D3G
- Entrez Gene ID
-
101060321
- Gene Name
- TBC1 domain family member 3G
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 36322242 - 36334759 on Build GRCh38
- Gene Symbol
- TBC1D3H
- Entrez Gene ID
-
729877
- Gene Name
- TBC1 domain family member 3H
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 36377475 - 36388448 on Build GRCh38
- Gene Symbol
- TBC1D3I
- Entrez Gene ID
-
102724862
- Gene Name
- TBC1 domain family member 3I
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 36253639 - 36264601 on Build GRCh38
- Gene Symbol
- TBC1D3K
- Entrez Gene ID
-
101060351
- Gene Name
- TBC1 domain family member 3K
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 37923805 - 37935371 on Build GRCh38
- Gene Symbol
- TBC1D3L
- Entrez Gene ID
-
101060376
- Gene Name
- TBC1 domain family member 3L
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 37978155 - 37990691 on Build GRCh38
View Full Product Details