Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COLQ
Entrez Gene ID
8292
Gene Name
collagen like tail subunit of asymmetric acetylcholinesterase
Gene Aliases
CMS5, EAD
Chromosome Location
Chr.3: 15450133 - 15521751 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080538.2 NP_536799.1 1 - 2 361 76
GenBank mRNA AK128401.1 - 1 - 2 361 76

View Full Product Details