Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COLQ
Entrez Gene ID
8292
Gene Name
collagen like tail subunit of asymmetric acetylcholinesterase
Gene Aliases
CMS5, EAD
Chromosome Location
Chr.3: 15450133 - 15521751 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005677.3 NP_005668.2 5 - 6 522 85
NM_080538.2 NP_536799.1 5 - 6 651 85
NM_080539.3 NP_536800.2 4 - 5 420 85
GenBank mRNA AF057036.1 - 5 - 6 522 85
AJ225895.1 - 5 - 6 515 85
AK128401.1 - 5 - 6 651 85
AK289688.1 - 5 - 6 541 85
AY150334.1 - 5 - 6 499 85
AY150335.1 - 5 - 6 499 85
AY150336.1 - 5 - 6 499 85
AY150337.1 - 5 - 6 499 85
AY150338.1 - 5 - 6 499 85
AY150339.1 - 5 - 6 499 85
BC074828.2 - 5 - 6 426 85
BC074829.2 - 5 - 6 426 85
BX500613.1 - 4 - 5 407 85

View Full Product Details