Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STRA13
Entrez Gene ID
201254
Gene Name
stimulated by retinoic acid 13
Gene Aliases
CENP-X, CENPX, D9, FAAP10, MHF2
Chromosome Location
Chr.17: 82018703 - 82022930 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271007.1 NP_001257936.1 1 - 2 84 88
NM_144998.3 NP_659435.2 1 - 2 84 88
XM_017024329.1 XP_016879818.1 1 - 2 105 88
GenBank mRNA BC009571.1 - 1 - 2 53 88
BC011610.1 - 1 - 2 35 88
U95006.1 - 1 - 2 39 88
U95007.1 - 1 - 2 39 88

View Full Product Details