The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145285.2 | NP_660328.2 | 1 - 2 | 558 | 95 |
GenBank mRNA | AK300856.1 | - | 1 - 2 | 510 | 95 |
AK313485.1 | - | 1 - 2 | 530 | 95 | |
BC025788.1 | - | 1 - 2 | 538 | 95 | |
BI764885.1 | - | 1 - 2 | 557 | 95 |