The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032140.1 | NP_115516.1 | 3 - 4 | 733 | 71 |
GenBank mRNA | AL136786.1 | - | 3 - 4 | 733 | 71 |
BC008284.1 | - | 3 - 4 | 700 | 71 |