Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GTPBP3
Entrez Gene ID
84705
Gene Name
GTP binding protein 3 (mitochondrial)
Gene Aliases
COXPD23, GTPBG3, MSS1, MTGP1, THDF1
Chromosome Location
Chr.19: 17334982 - 17342731 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128855.2 NP_001122327.1 2 - 3 403 80
NM_001195422.1 NP_001182351.1 2 - 3 396 80
NM_032620.3 NP_116009.2 2 - 3 403 80
NM_133644.3 NP_598399.2 2 - 3 403 80
GenBank mRNA AF360742.1 - 2 - 3 365 80
AF360743.1 - 2 - 3 365 80
AF360744.1 - 2 - 3 345 80
AK027606.1 - 2 - 3 350 80
AK091035.1 - 2 - 3 1108 80
AK291929.1 - 2 - 3 347 80
AK297953.1 - 2 - 3 378 80
AK298485.1 - - 361 80
AK311442.1 - 2 - 3 466 80
AY078987.1 - 2 - 3 365 80
AY078988.1 - 2 - 3 365 80
BC017207.2 - 2 - 3 335 80
BC019261.1 - 2 - 3 344 80
BI856784.1 - 2 - 3 373 80
DA666199.1 - 2 - 3 367 80
DC339966.1 - 2 - 3 458 80

View Full Product Details