The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_000029.3 | 2 - 3 | 220 | 92 | |
NR_024528.1 | 3 - 4 | 471 | 92 | ||
NR_024529.1 | 2 - 3 | 377 | 92 | ||
GenBank mRNA | BC000596.1 | - | 2 - 3 | 171 | 92 |
BC105600.1 | - | 2 - 3 | 172 | 92 | |
BG335018.1 | - | 3 - 4 | 415 | 92 | |
BI464848.1 | - | 2 - 3 | 381 | 92 |