Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FRMPD2
Entrez Gene ID
143162
Gene Name
FERM and PDZ domain containing 2
Gene Aliases
PDZD5C, PDZK4, PDZK5C
Chromosome Location
Chr.10: 48156559 - 48274898 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001018071.3 NP_001018081.3 27 - 28 3869 97
NM_001042512.2 NP_001035977.2 4 - 5 737 97
NM_001318191.1 NP_001305120.1 25 - 26 3510 97
XM_017015744.1 XP_016871233.1 4 - 5 553 97
GenBank mRNA AK123038.1 - 25 - 26 3510 97
AY491519.1 - 4 - 5 736 97
BC031614.1 - 6 - 7 732 97
BC073954.1 - 4 - 5 718 97
BC144636.1 - 4 - 5 678 97
BC144639.1 - 4 - 5 678 97
CR749241.1 - 27 - 28 3841 97

Gene Symbol
FRMPD2B
Entrez Gene ID
728798
Gene Name
FERM and PDZ domain containing 2B, pseudogene
Gene Aliases
FRMPD2L1, FRMPD2L2, FRMPD2P1, FRMPD2P2, FRMPDP2, PDZD5A, PDZD5B, PDZK5A, PDZK5B, bA556L1.2, yX59F3.2
Chromosome Location
Chr.10: 46876674 - 46895326 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_033172.1 4 - 5 791 97
GenBank mRNA AK294650.1 - 4 - 5 746 97
AK313659.1 - 4 - 5 748 97
AK315971.1 - 4 - 5 791 97
BC071635.1 - 4 - 5 703 97
BC072395.1 - 4 - 5 717 97
BC110798.1 - 4 - 5 714 97
BC137431.1 - 4 - 5 678 97
BC137434.1 - 4 - 5 678 97
BC137439.1 - 4 - 5 676 97
BC137440.1 - 4 - 5 678 97
BC144635.1 - 4 - 5 678 97

View Full Product Details