The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320373.1 | NP_001307302.1 | 2 - 3 | 196 | 70 |
NM_138345.2 | NP_612354.1 | 4 - 5 | 787 | 70 | |
XM_011513288.2 | XP_011511590.1 | 5 - 6 | 1423 | 70 | |
XM_011513289.2 | XP_011511591.1 | 5 - 6 | 1506 | 70 | |
XM_011513290.2 | XP_011511592.1 | 5 - 6 | 969 | 70 | |
XM_011513291.2 | XP_011511593.1 | 4 - 5 | 1635 | 70 | |
XM_011513292.2 | XP_011511594.1 | 4 - 5 | 1623 | 70 | |
GenBank mRNA | AK293885.1 | - | 2 - 3 | 196 | 70 |
BC136609.1 | - | 4 - 5 | 787 | 70 | |
DC308467.1 | - | 2 - 3 | 196 | 70 | |
DC310771.1 | - | 1 - 2 | 39 | 70 |