The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006605.4 | NP_001006606.2 | 4 - 5 | 566 | 95 |
NM_001252269.1 | NP_001239198.1 | 3 - 4 | 431 | 95 | |
NM_001252271.1 | NP_001239200.1 | 4 - 5 | 609 | 95 | |
GenBank mRNA | AK027146.1 | - | 1 - 2 | 127 | 95 |
AK303700.1 | - | 3 - 4 | 584 | 95 | |
AL536469.3 | - | 4 - 5 | 510 | 95 | |
DA155419.1 | - | 4 - 5 | 483 | 95 | |
DB199113.1 | - | 3 - 4 | 357 | 95 |