Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUDT19
Entrez Gene ID
390916
Gene Name
nudix hydrolase 19
Gene Aliases
RP2
Chromosome Location
Chr.19: 32691607 - 32713796 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105570.1 NP_001099040.1 2 - 3 921 122

View Full Product Details