The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011527471.2 | XP_011525773.1 | 14 - 15 | 1535 | 62 |
XM_011527472.2 | XP_011525774.1 | 14 - 15 | 1535 | 62 | |
XM_011527473.2 | XP_011525775.1 | 14 - 15 | 1535 | 62 | |
XM_011527474.2 | XP_011525776.1 | 13 - 14 | 1415 | 62 | |
XM_011527475.2 | XP_011525777.1 | 14 - 15 | 1535 | 62 | |
XM_011527476.2 | XP_011525778.1 | 14 - 15 | 1535 | 62 | |
XM_017027458.1 | XP_016882947.1 | 14 - 15 | 1535 | 62 | |
XM_017027459.1 | XP_016882948.1 | 4 - 5 | 518 | 62 | |
GenBank mRNA | BC049000.1 | - | 14 - 15 | 1775 | 62 |